Assay Design
Gene Transcripts
- Gene Symbol
- ARNT
- Entrez Gene ID
-
405
- Gene Name
- aryl hydrocarbon receptor nuclear translocator
- Gene Aliases
-
HIF-1-beta, HIF-1beta, HIF1-beta, HIF1B, HIF1BETA, TANGO, bHLHe2
- Chromosome Location
-
Chr.1: 150809705 - 150876768 on Build GRCh38
View Full Product Details