Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000409.4 | NP_000400.2 | 5 - 6 | 1113 | 81 |
NM_001319061.1 | NP_001305990.1 | 5 - 6 | 1484 | 81 | |
NM_001319062.1 | NP_001305991.1 | 4 - 5 | 937 | 81 | |
XM_006715073.3 | XP_006715136.1 | 4 - 5 | 326 | 81 | |
XM_011514537.2 | XP_011512839.1 | 5 - 6 | 1374 | 81 | |
XM_011514539.1 | XP_011512841.1 | 5 - 6 | 976 | 81 | |
GenBank mRNA | AK125780.1 | - | 5 - 6 | 1415 | 81 |
AK313865.1 | - | 4 - 5 | 908 | 81 | |
BC031663.1 | - | 5 - 6 | 1054 | 81 | |
L36859.1 | - | 3 - 4 | 462 | 81 |