Assay Design
Gene Transcripts
- Gene Symbol
- ATP6V1G2
- Entrez Gene ID
-
534
- Gene Name
- ATPase H+ transporting V1 subunit G2
- Gene Aliases
-
ATP6G, ATP6G2, NG38, VMA10
- Chromosome Location
-
Chr.6: 31544451 - 31546848 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_138282.2 |
NP_612139.1 |
1 - 2 |
222 |
66 |
GenBank mRNA |
BP195978.1 |
- |
1 - 2 |
129 |
66 |
View Full Product Details