Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130773.3 | NP_570129.1 | 2 - 3 | 552 | 72 |
XM_006712258.1 | XP_006712321.1 | 2 - 3 | 552 | 72 | |
XM_017003316.1 | XP_016858805.1 | 2 - 3 | 552 | 72 | |
GenBank mRNA | AB077881.1 | - | 2 - 3 | 188 | 72 |
AK056528.1 | - | 2 - 3 | 552 | 72 |