Assay Design



Gene Transcripts

Gene Symbol
CNTNAP5
Entrez Gene ID
129684
Gene Name
contactin associated protein like 5
Gene Aliases
caspr5
Chromosome Location
Chr.2: 124025287 - 124921201 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130773.3 NP_570129.1 17 - 18 3114 67
XM_006712258.1 XP_006712321.1 17 - 18 3117 67
GenBank mRNA AB077881.1 - 17 - 18 2750 67
AK056528.1 - 17 - 18 3117 67

View Full Product Details