Assay Design



Gene Transcripts

Gene Symbol
CNTNAP5
Entrez Gene ID
129684
Gene Name
contactin associated protein like 5
Gene Aliases
caspr5
Chromosome Location
Chr.2: 124025287 - 124921201 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130773.3 NP_570129.1 19 - 20 3576 97
XM_006712258.1 XP_006712321.1 19 - 20 3579 97
XM_017003316.1 XP_016858805.1 18 - 19 3339 97
GenBank mRNA AB077881.1 - 19 - 20 3212 97

View Full Product Details