Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244871.1 | NP_001231800.1 | 5 - 6 | 996 | 146 |
NM_001343.3 | NP_001334.2 | 5 - 6 | 996 | 146 | |
GenBank mRNA | AF188298.1 | - | 4 - 5 | 553 | 146 |
AK297984.1 | - | 1 - 2 | 110 | 146 | |
AK314381.1 | - | 5 - 6 | 744 | 146 | |
BC003064.2 | - | 5 - 6 | 592 | 146 | |
L16886.1 | - | 3 - 4 | 336 | 146 | |
U39050.1 | - | 4 - 5 | 564 | 146 | |
U53446.1 | - | 5 - 6 | 609 | 146 |