Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135099.1 | NP_001128571.1 | 4 - 5 | 495 | 55 |
NM_005656.3 | NP_005647.3 | 4 - 5 | 457 | 55 | |
XM_011529731.2 | XP_011528033.1 | 4 - 5 | 552 | 55 | |
XM_011529732.2 | XP_011528034.1 | 3 - 4 | 379 | 55 | |
XM_011529733.1 | XP_011528035.1 | 2 - 3 | 156 | 55 | |
GenBank mRNA | AF123453.1 | - | 2 - 3 | 323 | 55 |
AF270487.1 | - | 4 - 5 | 436 | 55 | |
AF329454.1 | - | 4 - 5 | 450 | 55 | |
AK222784.1 | - | 3 - 4 | 368 | 55 | |
AK291813.1 | - | 4 - 5 | 412 | 55 | |
AK296860.1 | - | 4 - 5 | 495 | 55 | |
AK313338.1 | - | 5 - 6 | 757 | 55 | |
BC035623.1 | - | 4 - 5 | 439 | 55 | |
BC051839.1 | - | 4 - 5 | 428 | 55 | |
BP276431.1 | - | 4 - 5 | 457 | 55 | |
U75329.1 | - | 3 - 4 | 379 | 55 |