Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001112706.2 | NP_001106177.1 | 5 - 6 | 974 | 107 |
NM_033128.3 | NP_149119.1 | 3 - 4 | 299 | 107 | |
GenBank mRNA | AB067492.1 | - | 5 - 6 | 974 | 107 |
AF276507.1 | - | 5 - 6 | 764 | 107 | |
AK027778.1 | - | 3 - 4 | 299 | 107 | |
AK075123.1 | - | 5 - 6 | 833 | 107 | |
AK290363.1 | - | 5 - 6 | 867 | 107 | |
BC021090.1 | - | 5 - 6 | 813 | 107 |