Assay Design



Gene Transcripts

Gene Symbol
SCIN
Entrez Gene ID
85477
Gene Name
scinderin
Gene Aliases
-
Chromosome Location
Chr.7: 12570349 - 12653603 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112706.2 NP_001106177.1 6 - 7 1097 104
NM_033128.3 NP_149119.1 4 - 5 422 104
GenBank mRNA AB067492.1 - 6 - 7 1097 104
AF276507.1 - 6 - 7 887 104
AK027778.1 - 4 - 5 422 104
AK075123.1 - 6 - 7 956 104
AK290363.1 - 6 - 7 990 104
BC021090.1 - 6 - 7 936 104

View Full Product Details