Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015545.3 | NP_056360.2 | 4 - 5 | 984 | 63 |
GenBank mRNA | AB014532.1 | - | 4 - 5 | 892 | 63 |
AK001615.1 | - | 4 - 5 | 899 | 63 | |
AK291004.1 | - | 4 - 5 | 875 | 63 | |
BC003688.1 | - | 3 - 4 | 497 | 63 | |
BC080580.1 | - | 4 - 5 | 932 | 63 | |
BC103495.1 | - | 4 - 5 | 909 | 63 | |
BC103502.1 | - | 4 - 5 | 909 | 63 | |
BC103661.1 | - | 2 - 3 | 158 | 63 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198879.1 | NP_001185808.1 | 5 - 6 | 1017 | 63 |
GenBank mRNA | AK295912.1 | - | 5 - 6 | 980 | 63 |