Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271519.1 | NP_001258448.1 | 24 - 25 | 3030 | 67 |
NM_001322830.1 | NP_001309759.1 | 25 - 26 | 3042 | 67 | |
NM_001322835.1 | NP_001309764.1 | 24 - 25 | 3033 | 67 | |
NM_001322837.1 | NP_001309766.1 | 24 - 25 | 3033 | 67 | |
XM_005269776.3 | XP_005269833.1 | 23 - 24 | 3010 | 67 | |
XM_005269789.2 | XP_005269846.1 | 24 - 25 | 2954 | 67 | |
XM_006717826.2 | XP_006717889.1 | 23 - 24 | 2942 | 67 | |
XM_011539774.1 | XP_011538076.1 | 26 - 27 | 3241 | 67 | |
XM_011539777.1 | XP_011538079.1 | 25 - 26 | 3196 | 67 | |
XM_011539778.2 | XP_011538080.1 | 24 - 25 | 3116 | 67 | |
XM_011539785.1 | XP_011538087.1 | 24 - 25 | 3019 | 67 | |
XM_017016207.1 | XP_016871696.1 | 26 - 27 | 3328 | 67 | |
XM_017016208.1 | XP_016871697.1 | 26 - 27 | 3328 | 67 | |
XM_017016209.1 | XP_016871698.1 | 25 - 26 | 3316 | 67 | |
XM_017016212.1 | XP_016871701.1 | 25 - 26 | 3154 | 67 | |
XM_017016214.1 | XP_016871703.1 | 24 - 25 | 3142 | 67 | |
XM_017016215.1 | XP_016871704.1 | 25 - 26 | 3109 | 67 | |
XM_017016218.1 | XP_016871707.1 | 23 - 24 | 2479 | 67 | |
GenBank mRNA | AY445624.1 | - | 1 - 2 | 59 | 67 |
BC144496.1 | - | 24 - 25 | 2933 | 67 |