Assay Design



Gene Transcripts

Gene Symbol
PDCD2
Entrez Gene ID
5134
Gene Name
programmed cell death 2
Gene Aliases
RP8, ZMYND7
Chromosome Location
Chr.6: 170575572 - 170584692 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199461.1 NP_001186390.1 2 - 3 632 135
NM_001199462.1 NP_001186391.1 3 - 4 495 135
NM_001199463.1 NP_001186392.1 3 - 4 495 135
NM_001199464.1 NP_001186393.1 3 - 4 495 135
NM_002598.3 NP_002589.2 2 - 3 632 135
NM_144781.2 NP_659005.1 2 - 3 632 135
XM_017010926.1 XP_016866415.1 2 - 3 573 135
GenBank mRNA AJ420535.1 - 2 - 3 378 135
AK297343.1 - 3 - 4 495 135
AW966881.1 - 2 - 3 367 135
AY948416.1 - 3 - 4 422 135
AY948417.1 - 3 - 4 422 135
BC008378.1 - 2 - 3 594 135
BC067759.1 - 2 - 3 508 135
BX385363.2 - 3 - 4 455 135
S78085.1 - 2 - 3 550 135

View Full Product Details