Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199461.1 | NP_001186390.1 | 1 - 2 | 396 | 120 |
NM_001199462.1 | NP_001186391.1 | 2 - 3 | 259 | 120 | |
NM_001199463.1 | NP_001186392.1 | 2 - 3 | 259 | 120 | |
NM_001199464.1 | NP_001186393.1 | 2 - 3 | 259 | 120 | |
NM_002598.3 | NP_002589.2 | 1 - 2 | 396 | 120 | |
NM_144781.2 | NP_659005.1 | 1 - 2 | 396 | 120 | |
XM_017010926.1 | XP_016866415.1 | 1 - 2 | 337 | 120 | |
GenBank mRNA | AJ420535.1 | - | 1 - 2 | 142 | 120 |
AK297343.1 | - | 2 - 3 | 259 | 120 | |
AW966881.1 | - | 1 - 2 | 131 | 120 | |
AY948416.1 | - | 2 - 3 | 186 | 120 | |
AY948417.1 | - | 2 - 3 | 186 | 120 | |
BC008378.1 | - | 1 - 2 | 358 | 120 | |
BG721244.1 | - | 1 - 2 | 369 | 120 | |
BX385363.2 | - | 2 - 3 | 219 | 120 | |
DC351104.1 | - | 2 - 3 | 259 | 120 |