Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161707.1 | NP_001155179.1 | 9 - 10 | 1492 | 74 |
NM_001301097.1 | NP_001288026.1 | 9 - 10 | 1492 | 74 | |
NM_032531.3 | NP_115920.1 | 9 - 10 | 1492 | 74 | |
XM_011543026.2 | XP_011541328.1 | 9 - 10 | 1236 | 74 | |
XM_011543027.2 | XP_011541329.1 | 10 - 11 | 1697 | 74 | |
XM_011543028.2 | XP_011541330.1 | 10 - 11 | 1556 | 74 | |
XM_011543030.2 | XP_011541332.1 | 9 - 10 | 1233 | 74 | |
XM_011543031.2 | XP_011541333.1 | 10 - 11 | 1691 | 74 | |
XM_011543032.2 | XP_011541334.1 | 9 - 10 | 1233 | 74 | |
XM_011543033.1 | XP_011541335.1 | 10 - 11 | 1697 | 74 | |
XM_017018418.1 | XP_016873907.1 | 9 - 10 | 1787 | 74 | |
XM_017018419.1 | XP_016873908.1 | 10 - 11 | 1697 | 74 | |
XM_017018420.1 | XP_016873909.1 | 10 - 11 | 1556 | 74 | |
GenBank mRNA | AB058770.1 | - | 9 - 10 | 1381 | 74 |
AF480410.1 | - | 9 - 10 | 1131 | 74 | |
AY358743.1 | - | 9 - 10 | 1432 | 74 | |
BC101775.1 | - | 9 - 10 | 1217 | 74 | |
BC101801.1 | - | 9 - 10 | 1217 | 74 |