Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320714.1 | NP_001307643.1 | 24 - 25 | 5505 | 112 |
NM_005128.3 | NP_005119.2 | 24 - 25 | 5396 | 112 | |
XM_017028509.1 | XP_016883998.1 | 21 - 22 | 4899 | 112 | |
GenBank mRNA | AB023150.2 | - | 21 - 22 | 4865 | 112 |
AJ237839.2 | - | 24 - 25 | 5402 | 112 | |
AK026478.1 | - | 3 - 4 | 332 | 112 | |
BC012791.2 | - | 6 - 7 | 1038 | 112 |