Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320714.1 | NP_001307643.1 | 29 - 30 | 6015 | 83 |
NM_005128.3 | NP_005119.2 | 29 - 30 | 5906 | 83 | |
XM_017028509.1 | XP_016883998.1 | 26 - 27 | 5409 | 83 | |
GenBank mRNA | AB023150.2 | - | 26 - 27 | 5375 | 83 |
AJ237839.2 | - | 29 - 30 | 5912 | 83 | |
AK026478.1 | - | 8 - 9 | 842 | 83 | |
BC012791.2 | - | 11 - 12 | 1527 | 83 |