Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320714.1 | NP_001307643.1 | 36 - 37 | 6832 | 97 |
NM_005128.3 | NP_005119.2 | 36 - 37 | 6723 | 97 | |
XM_017028509.1 | XP_016883998.1 | 33 - 34 | 6226 | 97 | |
GenBank mRNA | AB023150.2 | - | 33 - 34 | 6192 | 97 |
AJ237839.2 | - | 36 - 37 | 6729 | 97 | |
AK026478.1 | - | 15 - 16 | 1659 | 97 | |
BC012791.2 | - | 18 - 19 | 2344 | 97 |