Assay Design



Gene Transcripts

Gene Symbol
FAM83D
Entrez Gene ID
81610
Gene Name
family with sequence similarity 83 member D
Gene Aliases
C20orf129, CHICA, dJ616B8.3
Chromosome Location
Chr.20: 38926312 - 38953060 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030919.2 NP_112181.3 2 - 3 784 129
XM_017028088.1 XP_016883577.1 2 - 3 667 129
GenBank mRNA AK055793.1 - 2 - 3 786 129
BC001068.2 - 2 - 3 670 129
BC006553.2 - 2 - 3 670 129
BC053683.1 - 2 - 3 671 129
BC063661.1 - 2 - 3 670 129
BC080188.1 - 2 - 3 671 129

View Full Product Details