Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003129.3 | NP_003120.2 | 8 - 9 | 2274 | 101 |
XM_011517246.2 | XP_011515548.1 | 8 - 9 | 1372 | 101 | |
GenBank mRNA | AF098865.1 | - | 8 - 9 | 1503 | 101 |
AK225085.1 | - | 8 - 9 | 1564 | 101 | |
AK295302.1 | - | 8 - 9 | 1174 | 101 | |
AK313384.1 | - | 8 - 9 | 1392 | 101 | |
BC017033.1 | - | 8 - 9 | 1506 | 101 | |
BX647400.1 | - | 8 - 9 | 2208 | 101 | |
BX647605.1 | - | 8 - 9 | 2246 | 101 | |
D78129.1 | - | 6 - 7 | 792 | 101 |