Assay Design



Gene Transcripts

Gene Symbol
CNTNAP5
Entrez Gene ID
129684
Gene Name
contactin associated protein like 5
Gene Aliases
caspr5
Chromosome Location
Chr.2: 124025287 - 124921201 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006712258.1 XP_006712321.1 7 - 8 1429 65
XM_017003316.1 XP_016858805.1 7 - 8 1429 65
GenBank mRNA AK056528.1 - 7 - 8 1429 65

View Full Product Details