Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001202469.1 | NP_001189398.1 | 4 - 5 | 504 | 87 |
NM_001483.2 | NP_001474.1 | 6 - 7 | 621 | 87 | |
GenBank mRNA | AF029786.1 | - | 6 - 7 | 596 | 87 |
AJ001259.1 | - | 6 - 7 | 588 | 87 | |
AK097049.1 | - | 4 - 5 | 490 | 87 | |
AK125036.1 | - | 3 - 4 | 2877 | 87 | |
AK315648.1 | - | 6 - 7 | 606 | 87 | |
BC000732.2 | - | 6 - 7 | 594 | 87 | |
BC001837.2 | - | 6 - 7 | 594 | 87 | |
BC030821.1 | - | 6 - 7 | 603 | 87 | |
BT007112.1 | - | 6 - 7 | 588 | 87 | |
CR407612.1 | - | 6 - 7 | 588 | 87 | |
CX166452.1 | - | 6 - 7 | 621 | 87 |