Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031745.4 | NP_001026915.1 | 5 - 6 | 782 | 57 |
XM_005261988.3 | XP_005262045.1 | 5 - 6 | 990 | 57 | |
GenBank mRNA | AK292871.1 | - | 5 - 6 | 737 | 57 |
BC036767.1 | - | 5 - 6 | 697 | 57 |