Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000426.3 | NP_000417.2 | 24 - 25 | 3659 | 88 |
NM_001079823.1 | NP_001073291.1 | 24 - 25 | 3659 | 88 | |
XM_005266981.3 | XP_005267038.1 | 25 - 26 | 3923 | 88 | |
XM_005266982.3 | XP_005267039.1 | 25 - 26 | 3923 | 88 | |
XM_011535820.2 | XP_011534122.1 | 25 - 26 | 3923 | 88 | |
XM_017010851.1 | XP_016866340.1 | 25 - 26 | 6311 | 88 | |
XM_017010852.1 | XP_016866341.1 | 14 - 15 | 5200 | 88 | |
XM_017010853.1 | XP_016866342.1 | 25 - 26 | 3923 | 88 | |
GenBank mRNA | DB106101.1 | - | 3 - 4 | 517 | 88 |
Z26653.1 | - | 24 - 25 | 3603 | 88 |