Assay Design



Gene Transcripts

Gene Symbol
LAMA2
Entrez Gene ID
3908
Gene Name
laminin subunit alpha 2
Gene Aliases
LAMM
Chromosome Location
Chr.6: 128883141 - 129516566 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000426.3 NP_000417.2 39 - 40 5833 79
NM_001079823.1 NP_001073291.1 39 - 40 5833 79
XM_005266981.3 XP_005267038.1 40 - 41 6097 79
XM_005266982.3 XP_005267039.1 40 - 41 6097 79
XM_011535820.2 XP_011534122.1 40 - 41 6097 79
XM_017010851.1 XP_016866340.1 40 - 41 8485 79
XM_017010852.1 XP_016866341.1 29 - 30 7374 79
GenBank mRNA AB208922.1 - 14 - 15 1935 79
Z26653.1 - 39 - 40 5777 79

View Full Product Details