Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000426.3 | NP_000417.2 | 50 - 51 | 7256 | 118 |
NM_001079823.1 | NP_001073291.1 | 50 - 51 | 7256 | 118 | |
XM_005266981.3 | XP_005267038.1 | 51 - 52 | 7520 | 118 | |
XM_005266982.3 | XP_005267039.1 | 51 - 52 | 7520 | 118 | |
XM_011535820.2 | XP_011534122.1 | 50 - 51 | 7514 | 118 | |
XM_017010851.1 | XP_016866340.1 | 51 - 52 | 9908 | 118 | |
XM_017010852.1 | XP_016866341.1 | 40 - 41 | 8797 | 118 | |
GenBank mRNA | AB208922.1 | - | 25 - 26 | 3358 | 118 |
M59832.1 | - | 10 - 11 | 1211 | 118 | |
Z26653.1 | - | 50 - 51 | 7200 | 118 |