Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000426.3 | NP_000417.2 | 4 - 5 | 738 | 113 |
NM_001079823.1 | NP_001073291.1 | 4 - 5 | 738 | 113 | |
XM_005266981.3 | XP_005267038.1 | 4 - 5 | 738 | 113 | |
XM_005266982.3 | XP_005267039.1 | 4 - 5 | 738 | 113 | |
XM_011535820.2 | XP_011534122.1 | 4 - 5 | 738 | 113 | |
XM_017010851.1 | XP_016866340.1 | 4 - 5 | 3126 | 113 | |
XM_017010853.1 | XP_016866342.1 | 4 - 5 | 738 | 113 | |
GenBank mRNA | Z26653.1 | - | 4 - 5 | 682 | 113 |