Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001305163.1 | NP_001292092.1 | 12 - 13 | 2304 | 115 |
NM_001305164.1 | NP_001292093.1 | 12 - 13 | 1683 | 115 | |
NM_133368.2 | NP_588609.1 | 12 - 13 | 1705 | 115 | |
XM_005256220.1 | XP_005256277.1 | 12 - 13 | 2012 | 115 | |
XM_011523427.1 | XP_011521729.1 | 13 - 14 | 1791 | 115 | |
XM_011523428.1 | XP_011521730.1 | 13 - 14 | 2105 | 115 | |
XM_011523430.1 | XP_011521732.1 | 13 - 14 | 1732 | 115 | |
XM_017023844.1 | XP_016879333.1 | 13 - 14 | 2478 | 115 | |
GenBank mRNA | AB075852.1 | - | 12 - 13 | 1625 | 115 |
AK027549.1 | - | 12 - 13 | 2255 | 115 | |
AK124601.1 | - | 4 - 5 | 2622 | 115 | |
AK172845.1 | - | 12 - 13 | 1661 | 115 | |
BC013173.2 | - | 12 - 13 | 1660 | 115 |