Assay Design



Gene Transcripts

Gene Symbol
RSPRY1
Entrez Gene ID
89970
Gene Name
ring finger and SPRY domain containing 1
Gene Aliases
SEMDFA
Chromosome Location
Chr.16: 57186284 - 57240475 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001305163.1 NP_001292092.1 1 - 2 767 136
GenBank mRNA AK027549.1 - 1 - 2 718 136

View Full Product Details