Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001305163.1 | NP_001292092.1 | 4 - 5 | 1445 | 111 |
NM_001305164.1 | NP_001292093.1 | 4 - 5 | 824 | 111 | |
NM_133368.2 | NP_588609.1 | 4 - 5 | 846 | 111 | |
XM_005256220.1 | XP_005256277.1 | 4 - 5 | 1153 | 111 | |
XM_011523427.1 | XP_011521729.1 | 5 - 6 | 932 | 111 | |
XM_011523428.1 | XP_011521730.1 | 5 - 6 | 1246 | 111 | |
XM_011523430.1 | XP_011521732.1 | 5 - 6 | 873 | 111 | |
XM_017023844.1 | XP_016879333.1 | 5 - 6 | 1619 | 111 | |
GenBank mRNA | AB075852.1 | - | 4 - 5 | 766 | 111 |
AK027549.1 | - | 4 - 5 | 1396 | 111 | |
AK172845.1 | - | 4 - 5 | 802 | 111 | |
BC013173.2 | - | 4 - 5 | 801 | 111 |