Assay Design



Gene Transcripts

Gene Symbol
RSPRY1
Entrez Gene ID
89970
Gene Name
ring finger and SPRY domain containing 1
Gene Aliases
SEMDFA
Chromosome Location
Chr.16: 57186284 - 57240475 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001305163.1 NP_001292092.1 6 - 7 1618 134
NM_001305164.1 NP_001292093.1 6 - 7 997 134
NM_133368.2 NP_588609.1 6 - 7 1019 134
XM_005256220.1 XP_005256277.1 6 - 7 1326 134
XM_011523427.1 XP_011521729.1 7 - 8 1105 134
XM_011523428.1 XP_011521730.1 7 - 8 1419 134
XM_011523430.1 XP_011521732.1 7 - 8 1046 134
XM_017023844.1 XP_016879333.1 7 - 8 1792 134
GenBank mRNA AB075852.1 - 6 - 7 939 134
AK027549.1 - 6 - 7 1569 134
AK172845.1 - 6 - 7 975 134
BC013173.2 - 6 - 7 974 134

View Full Product Details