Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001305163.1 | NP_001292092.1 | 9 - 10 | 1941 | 65 |
NM_001305164.1 | NP_001292093.1 | 9 - 10 | 1320 | 65 | |
NM_133368.2 | NP_588609.1 | 9 - 10 | 1342 | 65 | |
XM_005256220.1 | XP_005256277.1 | 9 - 10 | 1649 | 65 | |
XM_011523427.1 | XP_011521729.1 | 10 - 11 | 1428 | 65 | |
XM_011523428.1 | XP_011521730.1 | 10 - 11 | 1742 | 65 | |
XM_011523430.1 | XP_011521732.1 | 10 - 11 | 1369 | 65 | |
XM_017023844.1 | XP_016879333.1 | 10 - 11 | 2115 | 65 | |
GenBank mRNA | AB075852.1 | - | 9 - 10 | 1262 | 65 |
AK027549.1 | - | 9 - 10 | 1892 | 65 | |
AK124601.1 | - | 1 - 2 | 2259 | 65 | |
AK172845.1 | - | 9 - 10 | 1298 | 65 | |
BC013173.2 | - | 9 - 10 | 1297 | 65 |