Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193288.2 | NP_001180217.1 | 2 - 3 | 1124 | 72 |
NM_020344.3 | NP_065077.1 | 2 - 3 | 1124 | 72 | |
XM_005251425.2 | XP_005251482.1 | 3 - 4 | 1403 | 72 | |
XM_005251426.4 | XP_005251483.1 | 3 - 4 | 1935 | 72 | |
XM_006716750.3 | XP_006716813.1 | 3 - 4 | 1935 | 72 | |
XM_017014592.1 | XP_016870081.1 | 7 - 8 | 1874 | 72 | |
XM_017014593.1 | XP_016870082.1 | 3 - 4 | 1407 | 72 | |
GenBank mRNA | AF097366.1 | - | 2 - 3 | 72 | |
AF177987.1 | - | 2 - 3 | 72 | ||
BC069622.1 | - | 2 - 3 | 72 | ||
BC143889.1 | - | 2 - 3 | 72 |