Assay Design



Gene Transcripts

Gene Symbol
RBP5
Entrez Gene ID
83758
Gene Name
retinol binding protein 5
Gene Aliases
CRBP-III, CRBP3, CRBPIII, HRBPiso
Chromosome Location
Chr.12: 7119106 - 7130272 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017020002.1 XP_016875491.1 1 - 2 562 58

View Full Product Details