Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003565.2 | NP_003556.1 | 5 - 6 | 584 | 76 |
XM_011538798.2 | XP_011537100.1 | 5 - 6 | 361 | 76 | |
XM_011538799.2 | XP_011537101.1 | 5 - 6 | 361 | 76 | |
GenBank mRNA | AB018265.2 | - | 5 - 6 | 366 | 76 |
AF045458.1 | - | 5 - 6 | 584 | 76 |