Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001369.2 | NP_001360.1 | 11 - 12 | 1578 | 116 |
XM_005248262.3 | XP_005248319.2 | 11 - 12 | 1661 | 116 | |
XM_017009177.1 | XP_016864666.1 | 11 - 12 | 1661 | 116 | |
XM_017009178.1 | XP_016864667.1 | 11 - 12 | 2464 | 116 | |
XM_017009179.1 | XP_016864668.1 | 5 - 6 | 758 | 116 | |
XM_017009180.1 | XP_016864669.1 | 11 - 12 | 1661 | 116 | |
XM_017009181.1 | XP_016864670.1 | 11 - 12 | 1661 | 116 | |
XM_017009182.1 | XP_016864671.1 | 11 - 12 | 1661 | 116 | |
XM_017009183.1 | XP_016864672.1 | 11 - 12 | 1661 | 116 | |
XM_017009184.1 | XP_016864673.1 | 11 - 12 | 1661 | 116 | |
XM_017009187.1 | XP_016864676.1 | 11 - 12 | 1661 | 116 | |
GenBank mRNA | AK055138.1 | - | 11 - 12 | 1943 | 116 |
AY045575.1 | - | 11 - 12 | 1578 | 116 |