Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001369.2 | NP_001360.1 | 58 - 59 | 9938 | 68 |
XM_005248262.3 | XP_005248319.2 | 58 - 59 | 10021 | 68 | |
XM_017009177.1 | XP_016864666.1 | 58 - 59 | 10021 | 68 | |
XM_017009178.1 | XP_016864667.1 | 58 - 59 | 10824 | 68 | |
XM_017009179.1 | XP_016864668.1 | 52 - 53 | 9118 | 68 | |
XM_017009180.1 | XP_016864669.1 | 58 - 59 | 10021 | 68 | |
XM_017009181.1 | XP_016864670.1 | 58 - 59 | 10021 | 68 | |
XM_017009182.1 | XP_016864671.1 | 58 - 59 | 10021 | 68 | |
XM_017009185.1 | XP_016864674.1 | 30 - 31 | 5317 | 68 | |
XM_017009186.1 | XP_016864675.1 | 27 - 28 | 4733 | 68 | |
XM_017009188.1 | XP_016864677.1 | 24 - 25 | 5549 | 68 | |
GenBank mRNA | AB046823.2 | - | 6 - 7 | 1002 | 68 |
AB290164.1 | - | 11 - 12 | 1941 | 68 | |
AK128600.1 | - | 1 - 2 | 185 | 68 | |
AY045575.1 | - | 58 - 59 | 9938 | 68 |