Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017738.3 | NP_060208.2 | 19 - 20 | 3371 | 134 |
XM_005251492.1 | XP_005251549.1 | 19 - 20 | 3368 | 134 | |
XM_006716793.3 | XP_006716856.1 | 19 - 20 | 3169 | 134 | |
XM_017014839.1 | XP_016870328.1 | 19 - 20 | 3371 | 134 | |
XM_017014840.1 | XP_016870329.1 | 19 - 20 | 3371 | 134 | |
XM_017014841.1 | XP_016870330.1 | 18 - 19 | 3176 | 134 | |
XM_017014844.1 | XP_016870333.1 | 12 - 13 | 2047 | 134 | |
XM_017014845.1 | XP_016870334.1 | 10 - 11 | 2852 | 134 | |
XM_017014846.1 | XP_016870335.1 | 7 - 8 | 1375 | 134 | |
XM_017014847.1 | XP_016870336.1 | 7 - 8 | 1368 | 134 | |
GenBank mRNA | AK000283.1 | - | 15 - 16 | 2466 | 134 |
AK021596.1 | - | 7 - 8 | 1380 | 134 | |
AK308837.1 | - | 19 - 20 | 3323 | 134 | |
BX647069.1 | - | 19 - 20 | 3330 | 134 |