Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017738.3 | NP_060208.2 | 23 - 24 | 3936 | 101 |
XM_005251492.1 | XP_005251549.1 | 23 - 24 | 3933 | 101 | |
XM_006716793.3 | XP_006716856.1 | 23 - 24 | 3734 | 101 | |
XM_017014839.1 | XP_016870328.1 | 23 - 24 | 3936 | 101 | |
XM_017014840.1 | XP_016870329.1 | 23 - 24 | 3936 | 101 | |
XM_017014841.1 | XP_016870330.1 | 22 - 23 | 3741 | 101 | |
XM_017014844.1 | XP_016870333.1 | 16 - 17 | 2612 | 101 | |
XM_017014845.1 | XP_016870334.1 | 14 - 15 | 3417 | 101 | |
XM_017014846.1 | XP_016870335.1 | 11 - 12 | 1940 | 101 | |
XM_017014847.1 | XP_016870336.1 | 11 - 12 | 1933 | 101 | |
GenBank mRNA | AK000283.1 | - | 19 - 20 | 3031 | 101 |
AK021596.1 | - | 11 - 12 | 1945 | 101 | |
AK308837.1 | - | 23 - 24 | 3888 | 101 | |
BX647069.1 | - | 23 - 24 | 3895 | 101 |