Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017738.3 | NP_060208.2 | 8 - 9 | 1411 | 109 |
XM_005251492.1 | XP_005251549.1 | 8 - 9 | 1411 | 109 | |
XM_006716793.3 | XP_006716856.1 | 8 - 9 | 1209 | 109 | |
XM_011517941.2 | XP_011516243.1 | 8 - 9 | 1411 | 109 | |
XM_017014839.1 | XP_016870328.1 | 8 - 9 | 1411 | 109 | |
XM_017014840.1 | XP_016870329.1 | 8 - 9 | 1411 | 109 | |
XM_017014842.1 | XP_016870331.1 | 8 - 9 | 1411 | 109 | |
XM_017014843.1 | XP_016870332.1 | 8 - 9 | 1411 | 109 | |
GenBank mRNA | AK000283.1 | - | 4 - 5 | 506 | 109 |
AK308837.1 | - | 8 - 9 | 1366 | 109 | |
BX647069.1 | - | 8 - 9 | 1369 | 109 |