Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297614.2 | NP_001284543.1 | 18 - 19 | 2306 | 102 |
NM_001304479.1 | NP_001291408.1 | 19 - 20 | 2078 | 102 | |
NM_020468.5 | NP_065201.1 | 16 - 17 | 2174 | 102 | |
NM_153816.5 | NP_722523.1 | 19 - 20 | 2333 | 102 | |
XM_005248738.3 | XP_005248795.1 | 19 - 20 | 2306 | 102 | |
XM_005248740.3 | XP_005248797.1 | 17 - 18 | 2175 | 102 | |
XM_011535977.2 | XP_011534279.1 | 18 - 19 | 1749 | 102 | |
XM_011535979.1 | XP_011534281.1 | 17 - 18 | 1741 | 102 | |
XM_017011090.1 | XP_016866579.1 | 19 - 20 | 1815 | 102 | |
XM_017011091.1 | XP_016866580.1 | 18 - 19 | 2279 | 102 | |
XM_017011092.1 | XP_016866581.1 | 17 - 18 | 1943 | 102 | |
XM_017011093.1 | XP_016866582.1 | 19 - 20 | 2020 | 102 | |
XM_017011094.1 | XP_016866583.1 | 17 - 18 | 1915 | 102 | |
XM_017011095.1 | XP_016866584.1 | 17 - 18 | 1655 | 102 | |
XM_017011096.1 | XP_016866585.1 | 19 - 20 | 2044 | 102 | |
XM_017011097.1 | XP_016866586.1 | 19 - 20 | 2028 | 102 | |
XM_017011098.1 | XP_016866587.1 | 18 - 19 | 1946 | 102 | |
XM_017011099.1 | XP_016866588.1 | 17 - 18 | 1912 | 102 | |
XM_017011100.1 | XP_016866589.1 | 16 - 17 | 1636 | 102 | |
XM_017011101.1 | XP_016866590.1 | 17 - 18 | 1718 | 102 | |
GenBank mRNA | AF121863.1 | - | 2 - 3 | 179 | 102 |
AK000362.1 | - | 19 - 20 | 2022 | 102 | |
AK026479.1 | - | 17 - 18 | 2438 | 102 | |
AK095380.1 | - | 19 - 20 | 2502 | 102 | |
AK125063.1 | - | 15 - 16 | 2385 | 102 | |
AK226097.1 | - | 7 - 8 | 644 | 102 | |
AK295417.1 | - | 19 - 20 | 1807 | 102 | |
AY044865.1 | - | 16 - 17 | 1715 | 102 | |
BC005110.1 | - | 16 - 17 | 1903 | 102 | |
BC046520.2 | - | 18 - 19 | 1761 | 102 | |
BC068589.1 | - | 19 - 20 | 2072 | 102 | |
BC095419.1 | - | 19 - 20 | 1918 | 102 |