Assay Design



Gene Transcripts

Gene Symbol
SNX14
Entrez Gene ID
57231
Gene Name
sorting nexin 14
Gene Aliases
RGS-PX2, SCAR20
Chromosome Location
Chr.6: 85505496 - 85594156 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297614.2 NP_001284543.1 7 - 8 1066 111
NM_001304479.1 NP_001291408.1 7 - 8 811 111
NM_020468.5 NP_065201.1 5 - 6 934 111
NM_153816.5 NP_722523.1 7 - 8 1066 111
XM_005248738.3 XP_005248795.1 7 - 8 1039 111
XM_005248740.3 XP_005248797.1 5 - 6 908 111
XM_011535977.2 XP_011534279.1 6 - 7 482 111
XM_011535979.1 XP_011534281.1 5 - 6 474 111
XM_017011090.1 XP_016866579.1 7 - 8 548 111
XM_017011091.1 XP_016866580.1 7 - 8 1039 111
XM_017011092.1 XP_016866581.1 5 - 6 676 111
XM_017011093.1 XP_016866582.1 7 - 8 753 111
XM_017011094.1 XP_016866583.1 5 - 6 648 111
XM_017011096.1 XP_016866585.1 7 - 8 781 111
XM_017011097.1 XP_016866586.1 7 - 8 733 111
XM_017011099.1 XP_016866588.1 5 - 6 649 111
XM_017011100.1 XP_016866589.1 4 - 5 341 111
XM_017011101.1 XP_016866590.1 6 - 7 482 111
GenBank mRNA AK000362.1 - 7 - 8 755 111
AK026479.1 - 6 - 7 1198 111
AK095380.1 - 8 - 9 1265 111
AK125063.1 - 3 - 4 1118 111
AK295417.1 - 7 - 8 540 111
AY044865.1 - 5 - 6 475 111
BC005110.1 - 5 - 6 663 111
BC046520.2 - 7 - 8 521 111
BC068589.1 - 7 - 8 805 111
BC095419.1 - 7 - 8 651 111

View Full Product Details