Assay Design



Gene Transcripts

Gene Symbol
SORBS2
Entrez Gene ID
8470
Gene Name
sorbin and SH3 domain containing 2
Gene Aliases
ARGBP2, PRO0618
Chromosome Location
Chr.4: 185585444 - 185956716 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145670.1 NP_001139142.1 14 - 15 2034 90
NM_001145671.2 NP_001139143.1 16 - 17 1823 90
NM_001145672.1 NP_001139144.1 14 - 15 1461 90
NM_001145673.1 NP_001139145.1 16 - 17 2347 90
NM_001145674.1 NP_001139146.1 8 - 9 2634 90
NM_001145675.1 NP_001139147.1 8 - 9 1098 90
NM_001270771.1 NP_001257700.1 17 - 18 3669 90
NM_003603.6 NP_003594.3 16 - 17 2062 90
NM_021069.4 NP_066547.1 14 - 15 3449 90
XM_005263307.1 XP_005263364.1 18 - 19 2941 90
XM_005263308.1 XP_005263365.1 17 - 18 2299 90
XM_005263310.1 XP_005263367.1 17 - 18 2155 90
XM_005263311.1 XP_005263368.1 16 - 17 2140 90
XM_005263312.1 XP_005263369.1 16 - 17 2137 90
XM_005263313.1 XP_005263370.1 14 - 15 2038 90
XM_006714388.1 XP_006714451.1 17 - 18 2230 90
XM_006714390.1 XP_006714453.1 15 - 16 2047 90
XM_017008741.1 XP_016864230.1 20 - 21 3499 90
XM_017008748.1 XP_016864237.1 17 - 18 3199 90
XM_017008750.1 XP_016864239.1 20 - 21 3499 90
XM_017008751.1 XP_016864240.1 19 - 20 3415 90
XM_017008752.1 XP_016864241.1 19 - 20 3340 90
XM_017008753.1 XP_016864242.1 17 - 18 3199 90
XM_017008754.1 XP_016864243.1 16 - 17 3154 90
XM_017008755.1 XP_016864244.1 14 - 15 2164 90
XM_017008756.1 XP_016864245.1 13 - 14 1867 90
XM_017008757.1 XP_016864246.1 9 - 10 1269 90
XM_017008758.1 XP_016864247.1 17 - 18 2333 90
XM_017008759.1 XP_016864248.1 14 - 15 2760 90
XM_017008760.1 XP_016864249.1 16 - 17 2206 90
XM_017008762.1 XP_016864251.1 15 - 16 2083 90
XM_017008763.1 XP_016864252.1 14 - 15 2033 90
XM_017008764.1 XP_016864253.1 15 - 16 2014 90
XM_017008765.1 XP_016864254.1 16 - 17 2317 90
XM_017008766.1 XP_016864255.1 14 - 15 1990 90
XM_017008767.1 XP_016864256.1 15 - 16 2293 90
XM_017008768.1 XP_016864257.1 14 - 15 2211 90
XM_017008769.1 XP_016864258.1 14 - 15 1969 90
XM_017008770.1 XP_016864259.1 13 - 14 1945 90
XM_017008771.1 XP_016864260.1 15 - 16 2087 90
XM_017008772.1 XP_016864261.1 14 - 15 1921 90
XM_017008773.1 XP_016864262.1 15 - 16 2224 90
XM_017008774.1 XP_016864263.1 10 - 11 1714 90
XM_017008775.1 XP_016864264.1 19 - 20 3454 90
GenBank mRNA AB018320.1 - 14 - 15 3101 90
AF049884.1 - 16 - 17 2026 90
AF049885.1 - 15 - 16 1466 90
AK056628.1 - 17 - 18 3287 90
AK225327.1 - 14 - 15 2000 90
AK225812.1 - 16 - 17 2347 90
AK293400.1 - 14 - 15 1461 90
AK296461.1 - 8 - 9 2411 90
AK304653.1 - 8 - 9 834 90
BC011883.1 - 16 - 17 1790 90

View Full Product Details