Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020964.2 | NP_066015.2 | 27 - 28 | 4908 | 65 |
XM_011526122.1 | XP_011524424.1 | 27 - 28 | 4908 | 65 | |
XM_017025889.1 | XP_016881378.1 | 27 - 28 | 4890 | 65 | |
XM_017025890.1 | XP_016881379.1 | 27 - 28 | 4890 | 65 | |
XM_017025891.1 | XP_016881380.1 | 26 - 27 | 4767 | 65 | |
XM_017025892.1 | XP_016881381.1 | 26 - 27 | 4933 | 65 | |
XM_017025893.1 | XP_016881382.1 | 19 - 20 | 2780 | 65 | |
GenBank mRNA | AK023817.1 | - | 11 - 12 | 1587 | 65 |
BC130614.1 | - | 10 - 11 | 1441 | 65 | |
BC144104.1 | - | 10 - 11 | 1441 | 65 | |
BC144105.1 | - | 10 - 11 | 1441 | 65 |