Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020964.2 | NP_066015.2 | 29 - 30 | 5206 | 146 |
XM_011526122.1 | XP_011524424.1 | 29 - 30 | 5206 | 146 | |
XM_017025889.1 | XP_016881378.1 | 29 - 30 | 5188 | 146 | |
XM_017025890.1 | XP_016881379.1 | 29 - 30 | 5188 | 146 | |
XM_017025891.1 | XP_016881380.1 | 28 - 29 | 5065 | 146 | |
XM_017025892.1 | XP_016881381.1 | 28 - 29 | 5231 | 146 | |
XM_017025893.1 | XP_016881382.1 | 21 - 22 | 3078 | 146 | |
GenBank mRNA | AK023817.1 | - | 13 - 14 | 1885 | 146 |
BC130614.1 | - | 12 - 13 | 1739 | 146 | |
BC144104.1 | - | 12 - 13 | 1739 | 146 | |
BC144105.1 | - | 12 - 13 | 1739 | 146 | |
DB235183.1 | - | 1 - 2 | 89 | 146 |