Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020964.2 | NP_066015.2 | 18 - 19 | 3482 | 66 |
XM_011526122.1 | XP_011524424.1 | 18 - 19 | 3482 | 66 | |
XM_017025889.1 | XP_016881378.1 | 18 - 19 | 3464 | 66 | |
XM_017025890.1 | XP_016881379.1 | 18 - 19 | 3464 | 66 | |
XM_017025891.1 | XP_016881380.1 | 17 - 18 | 3341 | 66 | |
XM_017025892.1 | XP_016881381.1 | 17 - 18 | 3507 | 66 | |
XM_017025893.1 | XP_016881382.1 | 10 - 11 | 1354 | 66 | |
GenBank mRNA | AB046852.1 | - | 18 - 19 | 3417 | 66 |
AK023817.1 | - | 2 - 3 | 161 | 66 |