Product Details

Assay Reference Genome
Location

Chr.9:108868799 on build GRCh38
Cytoband
9q31.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
8518
Gene Symbol
IKBKAP
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein
Gene Aliases
DYS, ELP1, FD, IKAP, IKI3, TOT1
Location
Chr.9:108867520-108934332 on build GRCh38
Assay Gene Location
Within Exon 37
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
IKBKAP NM_001318360.1 37 4694 NP_001305289.1
NM_003640.4 37 4839 NP_003631.2
XM_011519136.1 37 4673 XP_011517438.1
XM_011519137.1 37 4447 XP_011517439.1
XM_017015238.1 35 4410 XP_016870727.1
AB210033.1 37 4601
AF044195.1 37 4618 AAC64258.1
AF153419.2 37 4625 AAG43369.1
AK001641.1 37 4623
AK022559.1 19 2308
AK024223.1 17 2082
AK123257.1 10 2395
AK127237.1 29 4254
AK225798.1 24 2923
AK289962.1 37 4631
AL049945.1 12 1539 CAB43219.1
BC033094.1 37 4401 AAH33094.1
CR749385.1 30 3666 CAH18235.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv6659 Chr9:108856263 - 108890282 on Build GRCh38 Insertion IKBKAP ACTL7A

View Full Product Details