Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002559.3 | NP_002550.2 | 6 - 7 | 748 | 82 |
XM_011545069.2 | XP_011543371.1 | 7 - 8 | 1409 | 82 | |
XM_011545070.2 | XP_011543372.1 | 7 - 8 | 715 | 82 | |
XM_011545071.2 | XP_011543373.1 | 6 - 7 | 1277 | 82 | |
XM_011545072.2 | XP_011543374.1 | 7 - 8 | 1541 | 82 | |
XM_011545073.1 | XP_011543375.1 | 4 - 5 | 265 | 82 | |
GenBank mRNA | AB016608.1 | - | 6 - 7 | 564 | 82 |
BC069162.1 | - | 6 - 7 | 640 | 82 | |
BC074792.2 | - | 6 - 7 | 595 | 82 | |
BC074793.2 | - | 6 - 7 | 595 | 82 | |
BC109287.2 | - | 6 - 7 | 598 | 82 |