Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021999.4 | NP_068839.1 | 4 - 5 | 793 | 127 |
GenBank mRNA | AF086918.1 | - | 4 - 5 | 685 | 127 |
AF092128.1 | - | 4 - 5 | 782 | 127 | |
AF136973.1 | - | 4 - 5 | 782 | 127 | |
AF152462.1 | - | 4 - 5 | 740 | 127 | |
AF246221.1 | - | 4 - 5 | 740 | 127 | |
AK000503.1 | - | 4 - 5 | 725 | 127 | |
AK027411.1 | - | 4 - 5 | 724 | 127 | |
AK130048.1 | - | 4 - 5 | 729 | 127 | |
BC000554.2 | - | 4 - 5 | 719 | 127 | |
BC016148.1 | - | 4 - 5 | 740 | 127 | |
BT006863.1 | - | 4 - 5 | 570 | 127 |