Assay Design
Gene Transcripts
- Gene Symbol
- RBFOX1
- Entrez Gene ID
-
54715
- Gene Name
- RNA binding protein, fox-1 homolog 1
- Gene Aliases
-
2BP1, A2BP1, FOX-1, FOX1, HRNBP1
- Chromosome Location
-
Chr.16: 5239749 - 7713343 on Build GRCh38
View Full Product Details